Assay Details
Gene Symbol: eif4ebp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212803.1 | NP_997968.1 | 2 - 3 | 462 | 96 | |
GenBank mRNA | BC066546.1 | - | 2 - 3 | 462 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
