Assay Details
Gene Symbol: mtrex | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214768.1 | NP_999933.1 | 4 - 5 | 393 | 93 | |
GenBank mRNA | BC061456.1 | - | 4 - 5 | 393 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
