Assay Details
Gene Symbol: ppid | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002065.1 | NP_001002065.1 | 3 - 4 | 424 | 153 | |
GenBank mRNA | BC071388.1 | - | 3 - 4 | 424 | 153 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
