Assay Details
Gene Symbol: asb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002736.1 | NP_001002736.1 | 3 - 4 | 605 | 133 | |
GenBank mRNA | BC076421.1 | - | 3 - 4 | 605 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
