Assay Details
Gene Symbol: ndufb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017880.1 | NP_001017880.1 | 3 - 4 | 405 | 136 | |
GenBank mRNA | BC092968.1 | - | 3 - 4 | 405 | 136 | |
BC154160.1 | - | 3 - 4 | 407 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
