Assay Details
Gene Symbol: ube2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037691.2 | NP_001032780.1 | 3 - 4 | 211 | 79 | |
XM_021476902.1 | XP_021332577.1 | 972 | 79 | |||
GenBank mRNA | BC107825.1 | - | 3 - 4 | 211 | 79 | |
EH441049.1 | - | 3 - 4 | 239 | 79 | ||
EH549222.1 | - | 3 - 4 | 242 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
