Assay Details
Gene Symbol: ube2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037691.2 | NP_001032780.1 | 6 - 7 | 559 | 90 | |
XM_021476902.1 | XP_021332577.1 | 1320 | 90 | |||
GenBank mRNA | BC107825.1 | - | 6 - 7 | 559 | 90 | |
EH441049.1 | - | 6 - 7 | 587 | 90 | ||
EH549222.1 | - | 6 - 7 | 590 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
