Assay Details
Gene Symbol: p4ha1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044998.1 | NP_001038463.1 | 12 - 13 | 1326 | 86 | |
XM_009306501.3 | XP_009304776.1 | 12 - 13 | 1529 | 86 | ||
XM_009306502.3 | XP_009304777.1 | 12 - 13 | 1529 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
