Assay Details
Gene Symbol: gna11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045036.2 | NP_001038501.1 | 2 - 3 | 321 | 87 | |
XM_005170474.4 | XP_005170531.1 | 2 - 3 | 403 | 87 | ||
XM_005170475.4 | XP_005170532.1 | 2 - 3 | 327 | 87 | ||
GenBank mRNA | BC162964.1 | - | 2 - 3 | 403 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
