Assay Details
Gene Symbol: znf395b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045115.1 | NP_001038580.1 | 3 - 4 | 342 | 67 | |
XM_009294770.3 | XP_009293045.1 | 3 - 4 | 818 | 67 | ||
GenBank mRNA | BC139507.1 | - | 2 - 3 | 271 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
