Assay Details
Gene Symbol: znf395b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045115.1 | NP_001038580.1 | 4 - 5 | 446 | 94 | |
XM_009294770.3 | XP_009293045.1 | 4 - 5 | 922 | 94 | ||
GenBank mRNA | BC139507.1 | - | 3 - 4 | 375 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
