Assay Details
Gene Symbol: znf395b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045115.1 | NP_001038580.1 | 5 - 6 | 649 | 87 | |
XM_009294770.3 | XP_009293045.1 | 5 - 6 | 1125 | 87 | ||
GenBank mRNA | BC139507.1 | - | 4 - 5 | 578 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
