Assay Details
Gene Symbol: znf395b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045115.1 | NP_001038580.1 | 6 - 7 | 753 | 105 | |
XM_009294770.3 | XP_009293045.1 | 6 - 7 | 1229 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
