Assay Details
Gene Symbol: znf395b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045115.1 | NP_001038580.1 | 8 - 9 | 1061 | 100 | |
XM_009294770.3 | XP_009293045.1 | 8 - 9 | 1537 | 100 | ||
GenBank mRNA | BC139507.1 | - | 7 - 8 | 990 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
