Assay Details
Gene Symbol: znf395b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045115.1 | NP_001038580.1 | 9 - 10 | 1184 | 74 | |
XM_009294770.3 | XP_009293045.1 | 9 - 10 | 1660 | 74 | ||
GenBank mRNA | BC139507.1 | - | 8 - 9 | 1113 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
