Assay Details
Gene Symbol: acsl3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045128.1 | NP_001038593.1 | 2 - 3 | 540 | 60 | |
XM_005159236.4 | XP_005159293.1 | 4 - 5 | 1048 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
