Assay Details
Gene Symbol: ero1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076638.1 | NP_001070106.1 | 11 - 12 | 875 | 95 | |
XM_005172944.4 | XP_005173001.1 | 11 - 12 | 1119 | 95 | ||
GenBank mRNA | BC124220.1 | - | 11 - 12 | 875 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
