Assay Details
Gene Symbol: bud23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082879.1 | NP_001076348.1 | 1 - 2 | 152 | 81 | |
GenBank mRNA | BC133941.1 | - | 1 - 2 | 152 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
