Assay Details
Gene Symbol: myo1ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100031.1 | NP_001093501.1 | 29 - 30 | 3037 | 73 | |
XM_005165347.4 | XP_005165404.1 | 29 - 30 | 3212 | 73 | ||
XM_005165349.4 | XP_005165406.1 | 29 - 30 | 3272 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
