Assay Details
Gene Symbol: chrnb3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173287.1 | NP_775394.1 | 4 - 5 | 368 | 64 | |
GenBank mRNA | AB087185.1 | - | 4 - 5 | 368 | 64 | |
BC163409.1 | - | 4 - 5 | 437 | 64 | ||
BC163411.1 | - | 4 - 5 | 437 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
