Assay Details
Gene Symbol: rpl13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198143.1 | NP_937786.1 | 2 - 3 | 249 | 82 | |
XM_005166666.4 | XP_005166723.1 | 3 - 4 | 394 | 82 | ||
GenBank mRNA | AF385081.1 | - | 2 - 3 | 249 | 82 | |
AY561516.1 | - | 3 - 4 | 293 | 82 | ||
BC075977.1 | - | 3 - 4 | 280 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
