Assay Details
Gene Symbol: atf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199723.1 | NP_956017.1 | 7 - 8 | 930 | 102 | |
GenBank mRNA | BC053252.1 | - | 7 - 8 | 930 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
