Assay Details
Gene Symbol: ints13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199883.1 | NP_956177.1 | 12 - 13 | 1684 | 94 | |
XM_017354950.1 | XP_017210439.1 | 13 - 14 | 1750 | 94 | ||
GenBank mRNA | BC045349.1 | - | 12 - 13 | 1684 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
