Assay Details
Gene Symbol: ints13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199883.1 | NP_956177.1 | 15 - 16 | 2217 | 90 | |
XM_017354950.1 | XP_017210439.1 | 16 - 17 | 2283 | 90 | ||
GenBank mRNA | BC045349.1 | - | 15 - 16 | 2217 | 90 | |
BC066530.1 | - | 15 - 16 | 2208 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
