Assay Details
Gene Symbol: ints13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199883.1 | NP_956177.1 | 2 - 3 | 497 | 117 | |
XM_017354950.1 | XP_017210439.1 | 2 - 3 | 539 | 117 | ||
GenBank mRNA | BC045349.1 | - | 2 - 3 | 497 | 117 | |
BC066530.1 | - | 2 - 3 | 488 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
