Assay Details
Gene Symbol: ints13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199883.1 | NP_956177.1 | 7 - 8 | 1083 | 68 | |
GenBank mRNA | BC045349.1 | - | 7 - 8 | 1083 | 68 | |
BC066530.1 | - | 7 - 8 | 1074 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
