Assay Details
Gene Symbol: unk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200236.2 | NP_956530.2 | 2 - 3 | 452 | 121 | |
GenBank mRNA | BC047170.1 | - | 2 - 3 | 452 | 121 | |
EH489030.1 | - | 2 - 3 | 401 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
