Assay Details
Gene Symbol: ptdss1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200956.1 | NP_957250.1 | 7 - 8 | 1036 | 89 | |
GenBank mRNA | BC044533.1 | - | 7 - 8 | 1036 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
