Assay Details
Gene Symbol: pgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213387.1 | NP_998552.1 | 7 - 8 | 822 | 83 | |
GenBank mRNA | BC046026.1 | - | 7 - 8 | 822 | 83 | |
BC065888.1 | - | 7 - 8 | 792 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
