Assay Details
Gene Symbol: cela1.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003426.1 | NP_001003426.1 | 1 - 2 | 69 | 91 | |
XM_021469316.1 | XP_021324991.1 | 1 - 2 | 110 | 91 | ||
GenBank mRNA | BC076223.1 | - | 1 - 2 | 69 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
