Assay Details
Gene Symbol: mrpl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013458.2 | NP_001013476.1 | 4 - 5 | 519 | 78 | |
GenBank mRNA | BC091546.1 | - | 4 - 5 | 520 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
