Assay Details
Gene Symbol: adss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017804.1 | NP_001017804.1 | 11 - 12 | 1174 | 96 | |
GenBank mRNA | BC092877.1 | - | 11 - 12 | 1174 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
