Assay Details
Gene Symbol: ripor2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045026.1 | NP_001038491.1 | 22 - 23 | 3220 | 72 | |
XM_005159634.4 | XP_005159691.1 | 21 - 22 | 3120 | 72 | ||
XM_009294297.3 | XP_009292572.1 | 21 - 22 | 2987 | 72 | ||
XM_021468090.1 | XP_021323765.1 | 21 - 22 | 3235 | 72 | ||
XM_021468091.1 | XP_021323766.1 | 21 - 22 | 3143 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
