Assay Details
Gene Symbol: tnikb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037676.2 | NP_001032765.2 | 11 - 12 | 1296 | 80 | |
XM_005162778.4 | XP_005162835.1 | 11 - 12 | 1326 | 80 | ||
XM_005162779.4 | XP_005162836.1 | 11 - 12 | 1325 | 80 | ||
XM_005162780.4 | XP_005162837.1 | 11 - 12 | 1325 | 80 | ||
XM_021470183.1 | XP_021325858.1 | 11 - 12 | 1326 | 80 | ||
XM_021470184.1 | XP_021325859.1 | 11 - 12 | 1326 | 80 | ||
XM_021470185.1 | XP_021325860.1 | 11 - 12 | 1326 | 80 | ||
XM_021470186.1 | XP_021325861.1 | 11 - 12 | 1327 | 80 | ||
XM_021470187.1 | XP_021325862.1 | 11 - 12 | 1325 | 80 | ||
XM_021470188.1 | XP_021325863.1 | 11 - 12 | 1325 | 80 | ||
GenBank mRNA | BC124125.1 | - | 11 - 12 | 1282 | 80 | |
BC165688.1 | - | 11 - 12 | 1053 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
