Assay Details
Gene Symbol: tnikb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037676.2 | NP_001032765.2 | 12 - 13 | 1486 | 87 | |
XM_005162778.4 | XP_005162835.1 | 12 - 13 | 1516 | 87 | ||
XM_005162779.4 | XP_005162836.1 | 12 - 13 | 1515 | 87 | ||
XM_005162780.4 | XP_005162837.1 | 12 - 13 | 1515 | 87 | ||
XM_021470183.1 | XP_021325858.1 | 12 - 13 | 1516 | 87 | ||
XM_021470184.1 | XP_021325859.1 | 12 - 13 | 1516 | 87 | ||
XM_021470185.1 | XP_021325860.1 | 12 - 13 | 1516 | 87 | ||
XM_021470186.1 | XP_021325861.1 | 12 - 13 | 1517 | 87 | ||
XM_021470187.1 | XP_021325862.1 | 12 - 13 | 1515 | 87 | ||
XM_021470188.1 | XP_021325863.1 | 12 - 13 | 1515 | 87 | ||
GenBank mRNA | BC124125.1 | - | 12 - 13 | 1472 | 87 | |
BC165688.1 | - | 12 - 13 | 1243 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
