Assay Details
Gene Symbol: dhrs12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076557.1 | NP_001070025.1 | 4 - 5 | 367 | 83 | |
GenBank mRNA | BC122377.1 | - | 4 - 5 | 367 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
