Assay Details
Gene Symbol: ubn2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080169.1 | NP_001073638.1 | 13 - 14 | 2162 | 85 | |
XM_005165715.4 | XP_005165772.1 | 13 - 14 | 2771 | 85 | ||
XM_017356493.2 | XP_017211982.1 | 12 - 13 | 2669 | 85 | ||
GenBank mRNA | BC129172.1 | - | 13 - 14 | 2162 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
