Assay Details
Gene Symbol: ubn2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080169.1 | NP_001073638.1 | 6 - 7 | 1506 | 68 | |
XM_005165715.4 | XP_005165772.1 | 6 - 7 | 2115 | 68 | ||
XM_017356493.2 | XP_017211982.1 | 5 - 6 | 2013 | 68 | ||
GenBank mRNA | BC129172.1 | - | 6 - 7 | 1506 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
