Assay Details
Gene Symbol: adat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083097.1 | NP_001076566.1 | 2 - 3 | 282 | 109 | |
XM_005160618.4 | XP_005160675.1 | 2 - 3 | 371 | 109 | ||
GenBank mRNA | BC135109.1 | - | 2 - 3 | 280 | 109 | |
BC160619.1 | - | 2 - 3 | 283 | 109 | ||
BC163783.1 | - | 2 - 3 | 109 | |||
BC163804.1 | - | 2 - 3 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
