Assay Details
Gene Symbol: hoxb3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131117.3 | NP_571192.2 | 4 - 5 | 1310 | 82 | |
XM_005171554.3 | XP_005171611.1 | 4 - 5 | 1018 | 82 | ||
XM_005171562.4 | XP_005171619.1 | 2 - 3 | 3382 | 82 | ||
XM_017353138.2 | XP_017208627.1 | 4 - 5 | 1780 | 82 | ||
XM_017353140.2 | XP_017208629.1 | 3 - 4 | 1429 | 82 | ||
XM_017353141.2 | XP_017208630.1 | 3 - 4 | 1009 | 82 | ||
XM_017353143.2 | XP_017208632.1 | 3 - 4 | 2595 | 82 | ||
XM_021469305.1 | XP_021324980.1 | 3 - 4 | 1022 | 82 | ||
XM_021469306.1 | XP_021324981.1 | 4 - 5 | 912 | 82 | ||
XM_021469307.1 | XP_021324982.1 | 3 - 4 | 949 | 82 | ||
GenBank mRNA | BC095559.1 | - | 4 - 5 | 1310 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
