Assay Details
Gene Symbol: nras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131145.2 | NP_571220.2 | 2 - 3 | 407 | 76 | |
GenBank mRNA | BC076419.1 | - | 2 - 3 | 386 | 76 | |
EH537868.1 | - | 2 - 3 | 446 | 76 | ||
U62619.1 | - | 2 - 3 | 396 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
