Assay Details
Gene Symbol: caspa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131505.2 | NP_571580.1 | 5 - 6 | 957 | 76 | |
GenBank mRNA | AF233434.1 | - | 5 - 6 | 936 | 76 | |
BC095022.1 | - | 5 - 6 | 957 | 76 | ||
BC164780.1 | - | 5 - 6 | 970 | 76 | ||
MG957992.1 | - | 5 - 6 | 936 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
