Assay Details
Gene Symbol: bmi1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194366.1 | NP_919347.1 | 7 - 8 | 814 | 64 | |
GenBank mRNA | AB074153.1 | - | 7 - 8 | 814 | 64 | |
BC049423.1 | - | 7 - 8 | 574 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
