Assay Details
Gene Symbol: col1a1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201478.1 | NP_958886.1 | 17 - 18 | 1210 | 78 | |
XM_005155976.4 | XP_005156033.1 | 17 - 18 | 1253 | 78 | ||
XM_005155980.4 | XP_005156037.1 | 17 - 18 | 1256 | 78 | ||
GenBank mRNA | BC058045.1 | - | 17 - 18 | 1210 | 78 | |
BC066384.1 | - | 17 - 18 | 1199 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
