Assay Details
Gene Symbol: gria4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212752.2 | NP_997917.2 | 15 - 16 | 2718 | 94 | |
XM_021468772.1 | XP_021324447.1 | 15 - 16 | 2789 | 94 | ||
GenBank mRNA | AF525748.2 | - | 15 - 16 | 2718 | 94 | |
BC162620.1 | - | 15 - 16 | 2657 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
