Assay Details
Gene Symbol: traf4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212817.1 | NP_997982.1 | 3 - 4 | 426 | 61 | |
GenBank mRNA | AJ556549.1 | - | 3 - 4 | 523 | 61 | |
BC078361.1 | - | 3 - 4 | 335 | 61 | ||
BC165750.1 | - | 3 - 4 | 334 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
