Assay Details
Gene Symbol: etfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212998.2 | NP_998163.1 | 3 - 4 | 386 | 135 | |
GenBank mRNA | BC045971.1 | - | 3 - 4 | 387 | 135 | |
BC152229.1 | - | 3 - 4 | 377 | 135 | ||
BC164332.1 | - | 3 - 4 | 404 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
