Assay Details
Gene Symbol: pik3r3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291329.1 | NP_001278258.1 | 6 - 7 | 819 | 139 | |
NM_213038.2 | NP_998203.2 | 5 - 6 | 982 | 139 | ||
GenBank mRNA | BC059572.1 | - | 5 - 6 | 963 | 139 | |
EH456435.1 | - | 6 - 7 | 853 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
