Assay Details
Gene Symbol: mapk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213473.1 | NP_998638.1 | 5 - 6 | 1031 | 112 | |
XM_005167152.4 | XP_005167209.1 | 5 - 6 | 1132 | 112 | ||
GenBank mRNA | BC044411.1 | - | 5 - 6 | 1031 | 112 | |
BC164564.1 | - | 3 - 4 | 885 | 112 | ||
DQ360073.1 | - | 3 - 4 | 851 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
