Assay Details
Gene Symbol: mapk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213473.1 | NP_998638.1 | 6 - 7 | 1239 | 107 | |
XM_005167152.4 | XP_005167209.1 | 6 - 7 | 1340 | 107 | ||
GenBank mRNA | BC044411.1 | - | 6 - 7 | 1239 | 107 | |
BC164564.1 | - | 4 - 5 | 1093 | 107 | ||
DQ360073.1 | - | 4 - 5 | 1059 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
